missing translation for 'onlineSavingsMsg'
Learn More

TTC30A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7211360 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16883935 20 μL 20µL
16873935 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16883935 Supplier Proteintech Supplier No. 253521AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Specifications

Antigen TTC30A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TTC30A
Gene Accession No. Q86WT1
Gene Alias IFT70, TPR repeat protein 30A, TTC30A
Gene Symbols TTC30A
Host Species Rabbit
Immunogen TTC30A Fusion Protein Ag18361
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100362431, 92104
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.