missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TST Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16311-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene is a mitochondrial matrix enzyme that is encoded by the nucleus. It may play roles in cyanide detoxification, the formation of iron-sulfur proteins, and the modification of sulfur-containing enzymes. The gene product contains two highly conservative domains (rhodanese homology domains), suggesting these domains have a common evolutionary origin.Specifications
| TST | |
| Polyclonal | |
| Unconjugated | |
| TST | |
| RDS, Rhodanese, Thiosulfate sulfurtransferase, TST | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22117, 25274, 7263 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24329, P52196, Q16762 | |
| TST | |
| TST Fusion Protein Ag9346 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction