Learn More
Prestige Medical TST Indicator Strips
Product Code. 14862041
Change view
Click to view available options
Quantity:
50 Pk.
Unit Size:
Pack of 50
Product Code. Quantity unitSize
14862041 50 Pk. Pack of 50
1 options
Product Code. 14862041
Brand: Prestige Medical 219277
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
TST Indicator Strips 126°C. Box of 50 strips.
  • TST Indicator Strips 126°C Box of 50 Strips
Specifications
Product Type Indicator Strip
Temperature (English) 258.8°F
Temperature (Metric) 126°C
Quantity 50 Pk.