Learn More
TST™ TST™ Control™ Self Adhesive Emulating Indicator Strip
Product Code. 12916996
Change view
Click to view available options
Temperature (Metric):
121°C to 134°C
134°C
Sterilization Time:
3.5 min.
5.3 and 15 min.
5.3 min.
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Sterilization Time Unit Size
12916996 134°C 3.5 min. Pack of 100
12906996 121°C to 134°C 5.3 min. Pack of 100
12926996 121°C to 134°C 5.3 and 15 min. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12916996 Supplier TST™ Supplier No. 2340AB
  • ONLINE EXCLUSIVE PRICE
    527.00 NOK
    444.55 NOK / Pack of 100
    Save 82.45 NOK
    15% Off
Estimated Shipment: 04-01-2027
Place the TST™ Control™ Self Adhesive Emulating Indicator Strip inside trays, packs or pouches when autoclaving so the indicator will confirm that good quality steam has penetrated to the point of placement.
  • ISO 11140 part 1, class 6 classification
  • Paper
  • Steam sterilization
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO 11140 part 1, class 6
For Use With (Application) For sterilization
For Use With (Equipment) Autoclave
Product Type Emulating Indicator Strip
Temperature (Metric) 134°C
Features When placed inside trays, packs or pouches, a TST Control Emulating Indicator will confirm that good quality steam has penetrated to the point of placement
Material Paper
Quantity 100 Pk.
Sterilization Indicator Yes
Sterilization Time 3.5 min.
Sterilization Method Steam