missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN32 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20320-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene, which is a member of the tetraspanin superfamily, is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of chromosome 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian and breast cancers. This gene is located among several imprinted genes; however, this gene, as well as the tumor-suppressing subchromosomal transferable fragment 4, escapes imprinting. This gene may play a role in maligcies and diseases that involve this region, and it is also involved in hematopoietic cell function. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
| TSPAN32 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN32 | |
| PHEMX, PHMX, Protein Phemx, tetraspanin 32, Tspan 32, TSPAN32, TSSC6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10077, 27027 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96QS1, Q9JHH2 | |
| TSPAN32 | |
| TSPAN32 Fusion Protein Ag14174 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction