missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20463-1-AP-150UL
This item is not returnable.
View return policy
Description
TSPAN2 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.Specifications
| TSPAN2 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN2 | |
| 6330415F13Rik, NET3, Tetraspan NET 3, tetraspanin 2, TSN2, TSPAN 2, TSPAN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10100, 64521, 70747 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60636, Q922J6, Q9JJW1 | |
| Tspan2 | |
| TSPAN2 Fusion Protein Ag14299 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction