Learn More
TSFM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11701-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 ).Specifications
TSFM | |
Polyclonal | |
Unconjugated | |
TSFM | |
COXPD3, EF TS, EF Tsmt, TSFM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10102 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P43897 | |
TSFM | |
TSFM Fusion Protein Ag2308 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |