Learn More
TSC22D1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10214-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against an internal region of human TSC22D1.
TSC22 domain family protein, or TSC22, is a transcription factor that belongs to the large family of early response genes. This transcriptional repressor is known to act on the C-type natriuretic peptide (CNP) promoter. TSC22 belongs to the TSC-22/Dip/Bun family and is an intracellular protein that may be found in the cytoplasm or nucleus. This transcription factor is known to regulate cell growth, differentiation and cell death and is involved in modulating the transcriptional activity of Smad3 and Smad4. This protein is ubiquitously expressed in most tissues and widely in both fetal and adult tissues. It is generally expressed in aortic endothelial cells, and induced by cytokines, including TGFB. These proteins may be possible therapeutic targets of leukemia and prostate cancer.Specifications
| TSC22D1 | |
| Polyclonal | |
| Unconjugated | |
| TSC22D1 | |
| Cerebral protein 2, KIAA1994, Regulatory protein TSC 22, RP11 269C23.2, TGFB1I4, TSC22, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22D1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 21807, 498545, 8848 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62500, P62501, Q15714 | |
| Tsc22d1 | |
| TSC22D1 Fusion Protein Ag0287 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |