Learn More
Thermo Scientific™ Oxoid™ Tryptose Cycloserine Agar (TSC)
Product Code. 15882457
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
Product Code. 15882457
Brand: Thermo Scientific™ PO5315A
Only null left
Add to basket
Add to basket
Ensure precise isolation of Clostridium species with this agar, offering enhanced selectivity for clinical and environmental samples.
Ensure precise isolation of Clostridium species with Tryptose Cycloserine Agar (TSC), offering enhanced selectivity for clinical and environmental samples.
TRUSTED_SUSTAINABILITY
Specifications
Color Straw
Content And Storage 2°C to 12°C
Format 90 mm Plate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17 g
Quantity 10 Plates