missing translation for 'onlineSavingsMsg'
Learn More

Scharlab Tryptic Soy Agar (TSA)

Product Code. 11481887
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481887 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11481887 Supplier Scharlab Supplier No. 01200500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Scharlab Tryptic Soy Agar is general purpose solid medium containing animal and plant peptone.

  • BAM/COMPF/EP/HP/ISO/USP
  • General purpose solid medium containing animal and plant peptone
  • According to Pharmacopoeial Harmonised Method and ISO 9308-1, 22717 and 2296 standards.
  • The medium preparated Art. No.: 064-PA0100 contains defibrinated sheep blood.

Specifications

Certifications/Compliance ISO 9308-1, 22717, 2296
Form Solid
Media Type Tryptic Soy Agar
Product Type Dehydrated Culture Media
Formulation Casein Peptone 15.0 g/L, Soy Peptone 5.0 g/L, Sodium Chloride 5.0 g/L, Agar 15.0 g/L
For Use With (Application) General Cell Culture
Incubation Conditions 30°C to 35°C
Incubation Time 24 to 48 hr.
pH 7.3 ±0.2
Quantity 500 g
Solubility Insoluble
Show More Show Less
compliance-icons
Product Identifier
  • Tryptic Soy agar
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Pyruvic acid sodiumsalt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.