missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPV2 Monoclonal Antibody (OTI2G10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813002
This item is not returnable.
View return policy
Description
TRPV2 Monoclonal antibody specifically detects TRPV2 in Human samples. It is validated for Western BlotSpecifications
| TRPV2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y5S1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 51393 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2G10 | |
| Unconjugated | |
| Trpv2 | |
| GRC; growth factor regulated channel; growth factor-regulated calcium channel; Osm-9-like TRP channel 2; OTRPC2; RP23-234K24.7; Sac2b; Stretch-activated channel 2B; transient receptor potential cation channel subfamily V member 2; transient receptor potential cation channel, subfamily V, member 2; Trpv2; Vanilloid receptor-like protein 1; VRL; Vrl1; VRL-1 | |
| Human recombinant protein fragment corresponding to amino acids 655-764 of human TRPV2 (NP_057197) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction