missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPS1 Monoclonal Antibody (OTI1D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
2832.96 NOK - 6331.68 NOK
Specifications
| Antigen | TRPS1 |
|---|---|
| Clone | OTI1D9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17547874
|
Origene Technologies
TA813143S |
30 μL |
2832.96 NOK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17527874
|
Origene Technologies
TA813143 |
100 μL |
6331.68 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRPS1 Monoclonal antibody specifically detects TRPS1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TRPS1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| AI115454; AI447310; D15Ertd586e; GC79; LGCR; transcriptional repressor GATA binding 1; trichorhinophalangeal syndrome I; trichorhinophalangeal syndrome I (human); trichorhinophalangeal syndrome I homolog; tricho-rhino-phalangeal syndrome type I protein; TRPS1; Zinc finger protein GC79; zinc finger transcription factor Trps1 | |
| Human recombinant protein fragment corresponding to amino acids 985-1281 of human TRPS1 (NP_054831) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| TRPS1 |
| OTI1D9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q925H1, Q9UHF7 | |
| 299897, 7227, 83925 | |
| IgG2a | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title