Learn More
Tromethamine, Multi-Compendial, U.S.P., J.T. Baker™
Product Code. p-5391315 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
500g
Product Code. 15588244
Brand: Avantor J.T.Baker 410201
Chemical Identifiers
CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
Specifications
Packaging Poly Bottle
Quantity 500 g
Formula Weight 121.14
Grade Multi-Compendial/USP
Chemical Name or Material Tromethamine