missing translation for 'onlineSavingsMsg'
Learn More

Troglitazone, MedChemExpress

Product Code. p-200096139
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16474057

Brand: Medchem Express HY50935

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Troglitazone is a PPARγ agonist, with EC50s of 550 nM and 780 nM for human and murine PPARγ receptor, respectively.

Troglitazone is a PPARγ agonist, with EC50s of 550 nM and 780 nM for human and murine PPARγ receptor, respectively.

Specifications

Chemical Name or Material Troglitazone
CAS 97322-87-7
Color White
Purity Grade Notes Research
Molecular Formula C24H27NO5S
Quantity 50 mg
Synonym CS-045
Solubility Information DMSO : 160 mg/mL (362.37 mM; Need ultrasonic)
SMILES O=C(SC1CC(C=C2)=CC=C2OCC3(C)OC(C(CC3)=C4C)=C(C)C(C)=C4O)NC1=O
Molecular Weight (g/mol) 441.54
Formula Weight 441.54
Percent Purity 96.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.