Learn More
Interscience™ Trocar for Broth Bags
Product Code. 16312236
Change view
Click to view available options
Autoclavable:
Autoclavable
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Autoclavable Unit Size
16312236 Autoclavable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16312236 Supplier Interscience™ Supplier No. 513020
  • ONLINE EXCLUSIVE PRICE
    1895.00 NOK
    1845.27 NOK / Pack of 1
    Save 49.73 NOK
    2% Off
Estimated Shipment: 19-01-2027
Trocar compatible with most tubings.
  • Can be used with all tubings except those with an inner diameter of 1.6 mm
Specifications
Product Type Trocar
Autoclavable Autoclavable
For Use With (Application) Drawing Fluid from Bag
For Use With (Equipment) DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump, FlexiPump Pro