missing translation for 'onlineSavingsMsg'
Learn More

TRMT6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16727-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16846454

  • 3828.53 NOK / 150µL
Estimated Shipment: 09-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to 56 kDa TRMT6 protein.

Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.
Specifications

Specifications

TRMT6
Polyclonal
Unconjugated
TRMT6
CGI 09, GCD10, Gcd10p, KIAA1153, TRM6, TRMT6, tRNA(m1A58)MTase subunit TRM6
Rabbit
Antigen Affinity Chromatography
RUO
311441, 51605, 66926
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UJA5
TRMT6
TRMT6 Fusion Protein Ag10176
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers