missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMT6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16727-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to 56 kDa TRMT6 protein.
Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.Specifications
| TRMT6 | |
| Polyclonal | |
| Unconjugated | |
| TRMT6 | |
| CGI 09, GCD10, Gcd10p, KIAA1153, TRM6, TRMT6, tRNA(m1A58)MTase subunit TRM6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 311441, 51605, 66926 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UJA5 | |
| TRMT6 | |
| TRMT6 Fusion Protein Ag10176 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction