Description
- Provides rapid and reliable results
- Shows three different colors for each full pH unit
- Specially equipped with a hydrophobic barrier between different indicators
- Length: 5 m
Specifications
Specifications
| pH Range | 1 to 11 |
| Format | Roll |
| For Use With (Application) | For most precise reading these papers show three different colors for each full pH unit |
| Graduations | 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11 |
| Includes | 1 roll, 5m length |
| Quantity | 1 Each |
| Type | pH Paper |
| Length (Metric) | 5 m |
| Width (Metric) | 10 mm |