missing translation for 'onlineSavingsMsg'
Learn More

TRIS, USP grade, 99.0 to 101.0%, MP Biomedicals™

Product Code. 11492481 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
This item is not returnable. View return policy

Product Code. 11492481

Brand: MP Biomedicals 0219560580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tris is excellent biochemical and biological buffer for all applications where ultra high purity is required

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Trometamol, Tris-[hydroxymethyl]aminomethane
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99.0 to 101.0%
Formula Weight 121.14
Grade USP
Chemical Name or Material TRIS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.