Learn More
Tris(trimethylsilyloxy)ethylene, 95%, Thermo Scientific™
Product Code. p-3779840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10259463 1 g Glass bottle 1g
10707022 5 g Glass bottle 5g
2 options
Product Code. 10259463
Brand: Thermo Scientific Acros 216010010
Request Bulk or Custom Format
Chemical Identifiers
CAS 69097-20-7
Molecular Formula C11H28O3Si3
Molecular Weight (g/mol) 292.60
MDL Number MFCD00011641
InChI Key FCZGHPGTZRTDNN-UHFFFAOYSA-N
Synonym tris trimethylsilyloxy ethylene, tris trimethylsiloxy ethylene, vinyl-2-ylidenetris oxy tris trimethylsilane, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy-3,6-dioxa-2,7-disilaoct-4-ene, 3,6-dioxa-2,7-disilaoct-4-ene, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy, acmc-1b3cp, tris trimethylsiloxy ethene, tris trimethylsiloxy-ethylene, tris-trimethylsiloxy ethylene
PubChem CID 553067
IUPAC Name 1,2-bis(trimethylsilyloxy)ethenoxy-trimethylsilane
SMILES C[Si](C)(C)OC=C(O[Si](C)(C)C)O[Si](C)(C)C
Specifications
Color Colorless to Yellow
Density 0.8850g/mL
Boiling Point 90.0°C (1.0 mmHg)
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3SiO]2C=CHOSi(CH3)3
Refractive Index 1.4190 to 1.4220
Quantity 1 g
Fieser 08,523; 09,512
Specific Gravity 0.885
Solubility Information Solubility in water: insoluble.
Formula Weight 292.59
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyloxy)ethylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tris(trimethylsilyloxy)ethylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.