Learn More
Tris(trimethylsilyl)silane), TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Tris(trimethylsilyl)silane |
| CAS | 1873-77-4 |
| Molecular Formula | C9H28Si4 |
| Quantity | 5 g |
| Synonym | 1,1,1,3,3,3-Hexamethyl-2-(trimethylsilyl)trisilane, TTMSS, Tris(trimethylsilyl)silane |
| InChI Formula | InChI=1S/C9H28Si4/c1-11(2,3)10(12(4,5)6)13(7,8)9/h10H,1-9H3 |
| SMILES | C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C |
| IUPAC Name | bis(trimethylsilyl)silyl-trimethylsilane |
| Molecular Weight (g/mol) | 248.66 |
| Formula Weight | 248.1268 |