Learn More
Tris-Glycine, 1X Solution (Electrophoresis), Fisher BioReagents
Product Code. 11443024
Change view
Click to view available options
Quantity:
20 L
Unit Size:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443024 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11443024 Supplier Fisher Bioreagents Supplier No. BP243920
 Restricted Item
Estimated Shipment: 05-04-2027
Add to basket
Tris-Glycine is commonly used as a buffer in native polyacrylamide gel electrophoresis.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tris-Glycine
Name Note 1X Solution
CAS 56-40-6, 77-86-1, 7732-18-5
pH 8.3 to 8.5 (1X soln.)
Quantity 20 L
DNase Not Detected
Filtered Through Filtered through a 0.2-micron filter.
Identification Pass Test
Packaging Poly CUBE
Protease Not Detected
Grade Electrophoresis