Learn More
Tris, 99 to 99.5%, MP Biomedicals™
Product Code. 11438361 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Unit Size:
10kg
1kg
5kg
Product Code. 11438361
Brand: MP Biomedicals 0215217605
Chemical Identifiers
CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
Specifications
Color White
Assay Percent Range 99 to 99.5%
Quantity 5 kg
Formula Weight 121.14
Physical Form Crystalline Powder
Chemical Name or Material Tris