Learn More
Tris-(1,3-dichloroisopropyl) phosphate Standard 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30319166
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319166 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30319166 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011137AL
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tris(2-chloro-1-(chloromethyl)ethyl) phosphate
CAS 13674-87-8
Analyte or Component Names Tris(2-chloro-1-(chloromethyl)ethyl) phosphate
Shipping Condition Ice pack (-20°C)
Molecular Formula C9 H15 Cl6 O4 P
Concentration 50 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C9H15Cl6O4P/c10-1-7(2-11)17-20(16,18-8(3-12)4-13)19-9(5-14)6-15/h7-9H,1-6H2
SMILES ClCC(CCl)OP(=O)(OC(CCl)CCl)OC(CCl)CCl
IUPAC Name tris(1,3-dichloropropan-2-yl) phosphate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 430.9
Quantity 5 mL
Formula Weight 427.8839 g/mol