Learn More
Bochem™ Tripod
Product Code. 10048501
Change view
Click to view available options
Length (Metric):
210 mm
240 mm
Includes:
Screwed Feet
Welded Feet
Unit Size:
Each
Product Code. Length (Metric) Includes unitSize
10048501 210 mm Screwed Feet Each
15391961 240 mm Welded Feet Each
2 options
Product Code. 10048501
Brand: Bochem™ 5231
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bochem™ Tripod available with welded feet or screwed feet.
  • Welded feet on Stainless Steel
  • Screwed feet on Tempered Cast Iron
Specifications
Material Tempered Cast Iron
Length (Metric) 210 mm
Diameter (Metric) Inner 120 mm
Includes Screwed Feet
Product Type Tripod