Learn More
Bochem™ Tripod
Click to view available options
Length (Metric):
210 mm
240 mm
Includes:
Screwed Feet
Welded Feet
Unit Size:
Each
Description
- Welded feet on Stainless Steel
- Screwed feet on Tempered Cast Iron
Specifications
Specifications
| Material | Stainless Steel (18/10) |
| Length (Metric) | 240 mm |
| Diameter (Metric) Inner | 160 mm |
| Includes | Welded Feet |
| Product Type | Tripod |