Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringe
Shop alle Thermo Scientific produkter
Klik for at se tilgængelige muligheder
Volume (Metric):
10 μL
100 μL
250 μL
1 mL
Needle Gauge:
22 G
23 G
26s G
Pakningsstørrelse:
Each
Beskrivelse
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
- Borosilicate glass with metal flanges
- Needles and plungers are stainless steel
- Field-proven syringe design
Tekniske data
Tekniske data
| Material | Stainless Steel (Needle), Stainless Steel (Plunger) |
| Description | Sample Dilution |
| Needle Gauge | 22 G |
| Product Type | Gas Tight GC Autosampler Syringe, Fixed Needle |
| Volume (Metric) | 1 mL |
| Product Line | TriPlus |
| Length (Metric) Needle | 57 mm |
| Needle Point Style | LC |
| For Use With (Equipment) | TriPlus RSH Autosampler |
| Connection Type | Fixed |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion