Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringe
Product Code. 12876036 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
100 μL
250 μL
1 mL
Needle Gauge:
22 G
23 G
26s G
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge unitSize
12876036 100 μL 26s G Each
13426886 1 mL 22 G Each
12816036 10 μL 26s G Each
13436876 250 μL 23 G Each
13456866 100 μL 23 G Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12876036 Supplier Thermo Scientific™ Supplier No. 365H2161
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Needle), Stainless Steel (Plunger)
Description LV Splitless
Needle Gauge 26s G
Product Type Gas Tight GC Autosampler Syringe, Fixed Needle
Volume (Metric) 100 μL
Product Line TriPlus
Length (Metric) Needle 57 mm
Needle Point Style Cone
For Use With (Equipment) TriPlus RSH Autosampler and TriPlus 100LS Autosampler
Connection Type Fixed