Learn More
Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar with Neutralizers, VHP indicator, and 2D Barcode
BIOPHARMA
Product Code. 17657745
Click to view available options
Format:
55 mm plates
85 mm plate
Unit Size:
Pack of 100
Product Code. 17657745
Brand: Thermo Scientific™ R111TW1
Carry out environmental monitoring in your most regulated environments using Triple Wrapped Plates, for passive and active air sampling and surface testing applications.
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP indicator, Lecithin, Polysorbate 80, Sodium Thiosulphate and L-Histidin and 2D Barcode.
Achieve confidence in your results and ensure optimum media growth assurance using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.
Three protective layers reduce risk of contamination:
  • Primary film with moisture control patch
  • Secondary film VHP barrier with immobilized desiccant sachet
  • Tertiary film dust cover
  • Conveniently sold in packs of 100 which are sealed for security
The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.
TRUSTED_SUSTAINABILITY
Specifications
Color Ivory, transparent
Description Thermo Scientific™ Sterile Triple wrapped Tryptic Soy Agar contact with neutralizers and VHP indicator
Format 55 mm plates
Product Type Microbiology Media
Fill Weight 15 grams (+/- 0.5)
Packaging Type No cardboard inner packaging, triple wrapped-irradiated, with VHP barrier wrap
Quantity 100/pk