missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Triple Sugar Iron Agar (TSI Agar)
Description
- AOAC/BAM/COMPF/DIN/EP/IDF/ISO/USP
- Differential medium for identification of enterobacteria, by fermentation three sugars (lactose, sucrose and glucose) and hydrogen sulfide production
- ISO 6579, 6785 and 10272 standards
Specifications
Specifications
| Certifications/Compliance | ISO 6579, 6785, 10272 |
| Form | Solid |
| Media Type | Triple Sugar Iron Agar |
| Product Type | Differential Medium |
| Target Organism Class | Gram-negative Bacteria |
| Formulation | Peptone 20.000 g/L, Meat Extract 3.000 g/L, Yeast Extract 3.000 g/L, Lactose 10.000 g/L, Sucrose 10.000 g/L, Dextrose 1.000 g/L, Sodium Chloride 5.000 g/L, Iron(III) Citrate 0.300 g/L, Sodium Thiosulphate 0.300 g/L, Phenol Red 0.024 g/L, Agar 12.000 g/L |
| For Use With (Application) | Identification of Enterobacteria |
| Incubation Conditions | 37°C ± 1.0°C |
| Incubation Time | 24 ±3 hr. |
| pH | 7.4 ±0.2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?