Learn More
Julabo™ Triple-Insulated Tubing for Baths
Brand: Julabo™ 8930214
Additional Details : Weight : 0.50000kg
Specifications
CF30, CF31, CF40, CF41 Cryo-compact circulator, F25-HE refrigerated/heating circulator | |
Triple Insulated Tubing | |
Metal |
-100°C to +350°C | |
M16 x 1 connector | |
3 m |