Learn More
Triphenylacetic acid, 99%
Product Code. 10103480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
Product Code. Quantity Packaging unitSize
10103480 10 g Glass bottle 10g
10477340 5 g Glass bottle 5g
2 options
Product Code. 10103480
Brand: Thermo Scientific Acros 140300100
Request Bulk or Custom Format
CAS: 595-91-5 | C20H16O2 | 288.35 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 595-91-5
Molecular Formula C20H16O2
Molecular Weight (g/mol) 288.35
MDL Number MFCD00004185
InChI Key DCYGAPKNVCQNOE-UHFFFAOYSA-N
Synonym triphenylacetic acid, acetic acid, triphenyl, triphenyl acetic acid, benzeneacetic acid, .alpha.,.alpha.-diphenyl, nsc 61, tri phenylacetic acid, pubchem9200, alpha-toluic acid, alpha,alpha-diphenyl, acmc-1arvb, maybridge1_006927
PubChem CID 68992
IUPAC Name 2,2,2-triphenylacetic acid
SMILES OC(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 268°C to 269°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C6H5)3CCO2H
Quantity 10 g
Beilstein 09, 712
Formula Weight 288.34
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Triphenylacetic acid, 99%