Learn More
Tripelennamine hydrochloride, MedChemExpress
Product Code. 16481856
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
200mg
500mg
5g
Product Code. Quantity unitSize
16481856 500 mg 500mg
16479827 5 g 5g
16431679 1 g 1g
16427306 100 mg 100mg
16411266 200 mg 200mg
5 options
Product Code. 16481856
Brand: Medchem Express HY17428
MedChemExpress Tripelennamine hydrochloride, an ethylenediamine derivative, is a potent histamine H1-receptor antagonist. Tripelennamine hydrochloride lessens the allergic response of the organism caused by histamine. Tripelennamine hydrochloride can be used for the research of rhinitis, conjunctivitis, and allergic and anaphylactic reactions.
Tripelennamine hydrochloride, an ethylenediamine derivative, is a potent histamine H1-receptor antagonist. Tripelennamine hydrochloride lessens the allergic response of the organism caused by histamine. Tripelennamine hydrochloride can be used for the research of rhinitis, conjunctivitis, and allergic and anaphylactic reactions.
Specifications
Chemical Name or Material Tripelennamine hydrochloride
CAS 154-69-8
Color White
Purity Grade Notes Research
Molecular Formula C16H22ClN3
Quantity 500 mg
Solubility Information H2O : 100 mg/mL (342.68 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (85.67 mM; Need ultrasonic)
SMILES CN(C)CCN(CC1=CC=CC=C1)C2=NC=CC=C2.[H]Cl
Molecular Weight (g/mol) 291.82
Formula Weight 291.82
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation