missing translation for 'onlineSavingsMsg'
Learn More

Triolein, ≥95%, MP Biomedicals™

Product Code. 11403521 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403521 1 g 1g
11413521 5 g 5g
11423521 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403521 Supplier MP Biomedicals Supplier No. 0210312201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Triolein is used as a substrate for microbial lipases. Triolein is a symmetrical triglyceride

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-32-7
Molecular Formula C57H104O6
Molecular Weight (g/mol) 885.453
MDL Number MFCD00137563
InChI Key PHYFQTYBJUILEZ-BSCDBXJPSA-N
Synonym triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate
PubChem CID 45253964
IUPAC Name [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC

Specifications

Color Yellow
Density 0.91g/mL(lit.)
Assay Percent Range ≥95%
Quantity 1 g
Formula Weight 885.46
Physical Form Liquid
Chemical Name or Material Triolein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.