missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl isocyanate, TRC

Product Code. 30456389
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
Product Code. Quantity unitSize
30456389 5 g 5g
30455190 100 mg 100mg
30452539 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 30456389

Brand: Toronto Research Chemicals TRCT7981985G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-02-1
Formula Weight 115.045
InChI Formula InChI=1S/C4H9NOSi/c1-7(2,3)5-4-6/h1-3H3
IUPAC Name isocyanato(trimethyl)silane
Molecular Formula C4 H9 N O Si
Molecular Weight (g/mol) 115.206
SMILES C[Si](C)(C)N=C=O
Synonym Isocyanic Acid Trimethylsilyl Ester, Orgatix SI 130, Trimethylisocyanatosilane, Trimethylsilyl iIsocyanate
Chemical Name or Material Trimethylsilyl isocyanate

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.