missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl isocyanate, 94%

Product Code. p-7020615
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
11420473 10 g 10g
11430473 50 g 50g
11440473 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 11420473

Brand: Thermo Scientific Alfa Aesar A12633.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1118-02-1 | C4H9NOSi | 115.21 g/mol

Trimethylsilyl isocyanate is used in the preparation of 1-unsubstituted 4-(dialkylamino) imidazolin-2-ones. It finds application in the carbamoylation of aromatic hydrocarbons and alcohols. It is also used in the preparation of primary amides from Grignard reagents. Further, it is used in the synthesis of cyclic imides. It is also utilized for the synthesis of trifluoroacetyl isocyanate and cyclic imides. In addition to this, it is employed in the conversion of isocyanates to carbodiimides using cyclopentadienyl Mn(CO)3 catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl isocyanate is used in the preparation of 1-unsubstituted 4-(dialkylamino) imidazolin-2-ones. It finds application in the carbamoylation of aromatic hydrocarbons and alcohols. It is also used in the preparation of primary amides from Grignard reagents. Further, it is used in the synthesis of cyclic imides. It is also utilized for the synthesis of trifluoroacetyl isocyanate and cyclic imides. In addition to this, it is employed in the conversion of isocyanates to carbodiimides using cyclopentadienyl Mn(CO)3 catalysis.

Solubility
Miscible with ethers and o-dichlorobenzene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, strong bases, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-02-1
Molecular Formula C4H9NOSi
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001993
InChI Key NIZHERJWXFHGGU-UHFFFAOYSA-N
Synonym trimethylsilyl isocyanate, silane, isocyanatotrimethyl, trimethylsilylisocyanate, isocyanato trimethyl silane, trimethylisocyanatosilane, tmsisocyanate, tms isocyanate, tms-isocyanate, tms isocynate
PubChem CID 70696
IUPAC Name isocyanato(trimethyl)silane
SMILES C[Si](C)(C)N=C=O

Specifications

Melting Point -49°C
Density 0.859
Boiling Point 90°C to 92°C
Flash Point −2°C (28°F)
Odor Acrid
Refractive Index 1.396
Quantity 10 g
UN Number UN2478
Beilstein 1744962
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Miscible with ethers and o-dichlorobenzene.
Formula Weight 115.21
Percent Purity 94%
Chemical Name or Material Trimethylsilyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethylsilyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Trimethylsilyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.