Learn More
Trimethylsilyl acrylate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 13688-55-6 |
| Molecular Formula | C6H12O2Si |
| MDL Number | MFCD00156432 |
| Quantity | 25 g |
| SMILES | C=CC(O[Si](C)(C)C)=O |
| Molecular Weight (g/mol) | 144.24 |
| Percent Purity | 95% |
| Chemical Name or Material | Trimethylsilyl acrylate |