Learn More
Trimethyl Phosphate, Sigma-Aldrich™
Product Code. 15831441
Change view
Click to view available options
Quantity:
100 mL
2.5 L
Unit Size:
100mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15831441 2.5 L 2.5 L
15811441 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15831441 Supplier Sigma Aldrich Supplier No. 8211842500
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 512-56-1
Specifications
Melting Point -46°C
Density 1.21 g/cm3 at 20°C
Boiling Point 197°C/1013 hPa
Molecular Formula (CH3)3PO4
MDL Number MFCD00008348
Quantity 2.5 L
Synonym Trimethyl phosphate, Trimethyl phosphate
Molecular Weight (g/mol) 140.07
Grade Research