Learn More
Trimethoxyphenylsilane, Sigma-Aldrich™
Product Code. 32238960
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238960 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238960 Supplier Sigma Aldrich Supplier No. 8436580050
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
For synthesis
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material Trimethoxyphenylsilane
CAS 2996-92-1
Density 1.064 g/cm³
Flash Point 29°C (closed cup)
Molecular Formula (CH3O)3SiC6H5
Quantity 50 mL
Synonym Trimethoxyphenylsilane, Trimethoxyphenylsilane
Molecular Weight (g/mol) 198.29
Grade Not Available