Learn More
TRIM37 Rabbit Polyclonal Antibody, Proteintech
Product Code. p-200108264 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
30233251 20 μL 20µL
30235718 150 μL 150µL
2 options
Product Code. 30233251
Brand: Proteintech 130371AP20UL
Rabbit Polyclonal Antibody
The TRIM37 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRIM37. This antibody recognizes human, mouse antigen. The TRIM37 antibody has been validated for the following applications: WB, IHC, IF/ICC, IP, ELISA analysis.
Specifications
Antigen TRIM37
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA
Classification Polyclonal
Concentration 650 μg/mL
Conjugate Unconjugated
Gene Symbols KIAA0898; MUL; POB1; TEF3
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Antigen affinity purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4591
Target Species Human, Mouse
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 130 kDa
Product Type Polyclonal Antibody
Form Liquid
Isotype IgG