Learn More
TRIM3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28392-1-AP-150UL
 View more versions of this product
Product Code. 16836125
  • 4836.10 NOK / 150µL
Description
The TRIM3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRIM3. This antibody recognizes human, mouse, rat antigen. The TRIM3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TRIM3
Polyclonal
Unconjugated
150 μL
10612
IgG
Western Blot, ELISA
333 μg/mL
Rabbit
Primary
Human, Rat, Mouse