Learn More
TRIM3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28392-1-AP-150UL
Description
The TRIM3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRIM3. This antibody recognizes human, mouse, rat antigen. The TRIM3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TRIM3 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 10612 | |
| IgG |
| Western Blot, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |