missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20356-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic filaments. Its function has not been identified.Specifications
| TRIM2 | |
| Polyclonal | |
| Unconjugated | |
| TRIM2 | |
| KIAA0517, RING finger protein 86, RNF86, TRIM2, tripartite motif containing 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23321, 361970, 80890 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D3ZQG6, Q9C040, Q9ESN6 | |
| Trim2 | |
| TRIM2 Fusion Protein Ag14208 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction