missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl Borate, Sigma-Aldrich™

Product Code. 15854751
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15854751 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15854751 Supplier Sigma Aldrich Supplier No. 8412640100
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 5419-55-6
Melting Point -59°C
Density 0.82 g/cm3 at 20°C
Boiling Point 139°C to 141°C/1013 hPa
MDL Number MFCD00008872
Quantity 100 mL
Synonym Triisopropyl borate, Boric acid triisopropyl ester, Triisopropyl borate
Molecular Weight (g/mol) 188.07
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.