Learn More
Trigonelline chloride, MedChemExpress
Product Code. 16462376
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16462376
Brand: Medchem Express HYN0415
MedChemExpress Trigonelline chloride, an alkaloid with potential antidiabetic activity, is present in considerable amounts in coffee. Trigonelline chloride has anti-HSV-1 , antibacterial, and antifungal activities.
Trigonelline chloride, an alkaloid with potential antidiabetic activity, is present in considerable amounts in coffee. Trigonelline chloride has anti-HSV-1 , antibacterial, and antifungal activities.
Specifications
Chemical Name or Material Trigonelline chloride
Physical Form Solid
CAS 6138-41-6
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C7H8ClNO2
For Use With (Application) Cancer-programmed cell death
Synonym Trigonelline hydrochloride
Solubility Information DMSO : 125 mg/mL (720.05 mM; Need ultrasonic) ∣H2O : 100 mg/mL (576.04 mM; Need ultrasonic)
SMILES C[N+]1=CC(C(O)=O)=CC=C1.[Cl-]
Molecular Weight (g/mol) 173.6
Formula Weight 173.6
Percent Purity 98.22%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.