Learn More
Trifluoroacetic acid-D1, deuteration degree min. 99.5% for NMR spectroscopy MagniSolv™, MilliporeSigma™
Product Code. 15835670
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass Ampule
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15835670 10 mL Glass Ampule 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15835670 Supplier Sigma Aldrich Supplier No. 1133630010
 Restricted Item
Estimated Shipment: 26-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
For NMR Spectroscopy
TRUSTED_SUSTAINABILITY
Specifications
Physical Form Liquid
Odor Stinging
Packaging Glass Ampule
Quantity 10 mL
CAS 599-00-8
Assay Percent Range 99.5% min.
Molecular Formula C2HF3O2
Synonym trifluoroacetic acid-d, deuterotrifluoroacetic acid, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, acetic acid-d, 2,2,2-trifluoro, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d, tfa-d, cf3cood, trifluoroacetic 2h acid
InChI Key DTQVDTLACAAQTR-DYCDLGHISA-N
SMILES C(=O)(C(F)(F)F)O
IUPAC Name deuterio 2,2,2-trifluoroacetate
Molecular Weight (g/mol) 115.029
PubChem CID 71502
Formula Weight 115.03
Chemical Name or Material Trifluoroacetic acid-D1