Learn More
Triethylene Glycol, Sigma-Aldrich™
Product Code. 15829570
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829570 2.5 L 2.5 L
15819570 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15829570 Supplier Sigma Aldrich Supplier No. 8082452500
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 112-27-6
Specifications
Melting Point -7°C
Density 1.123 g/cm3 at 20°C
Boiling Point 285°C to 295°C/1013 hPa
Molecular Formula HO(CH2CH2O)3H
MDL Number MFCD00002880
Quantity 2.5 L
Synonym Triethylene glycol, Triglycol, Triethylene glycol
Molecular Weight (g/mol) 150.17
Grade Research