Learn More
Triethylamine Trishydrofluoride, Sigma-Aldrich™
Specifications
Specifications
| CAS | 73602-61-6 |
| Density | 1.00 g/cm3 at 20°C |
| Boiling Point | 70°C/19.9 hPa |
| Molecular Formula | (C2H5)3N · 3 HF |
| MDL Number | MFCD00043294 |
| Quantity | 10 mL |
| Synonym | Triethylamine trishydrofluoride, Triethylamine trishydrofluoride |
| Molecular Weight (g/mol) | 161.21 |
| Grade | Research |