Learn More
Triethyl Phosphate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 78-40-0 |
| Melting Point | -57°C |
| Density | 1.069 g/cm3 at 20°C |
| Boiling Point | 215°C/1013 hPa |
| Molecular Formula | (C2H5O)3PO |
| MDL Number | MFCD00009077 |
| Quantity | 1 L |
| Synonym | Triethyl phosphate, Phosphoric acid triethyl ester, TEP, Triethyl phosphate |
| Molecular Weight (g/mol) | 182.15 |
| Grade | Research |