Learn More
Triethyl Citrate, Sigma-Aldrich™
Product Code. 15888210
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15888210 500 mL 500mL
15878210 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15888210 Supplier Sigma Aldrich Supplier No. 8002510500
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product can replace highly toxic diluents in membrane preparation and thus aligns with Less Hazardous Chemical Syntheses.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 77-93-0
Specifications
Melting Point -55°C
Density 1.135 g/cm3 at 25°C
Boiling Point 294°C/1013 hPa
Molecular Formula HOC(COOC2H5)(CH2COOC2H5)2
MDL Number MFCD00009201
Quantity 500 mL
Synonym Triethyl citrate, Citric acid triethyl ester, Triethyl citrate
Molecular Weight (g/mol) 276.28
Grade Research