Learn More
Trichloro Bisphenol S, TRC
Product Code. 30452444
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452444 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30452444 Supplier Toronto Research Chemicals Supplier No. TRCT89686610MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trichloro Bisphenol S
CAS 80396-78-7
Molecular Formula C12H7Cl3O4S
Quantity 10 mg
Synonym 2,2',6-Trichloro-4,4'-sulfonyldiphenol, 2,6-Dichloro-4-((3-chloro-4-hydroxyphenyl)sulfonyl)phenol
InChI Formula InChI=1S/C12H7Cl3O4S/c13-8-3-6(1-2-11(8)16)20(18,19)7-4-9(14)12(17)10(15)5-7/h1-5,16-17H
SMILES Oc1ccc(cc1Cl)S(=O)(=O)c2cc(Cl)c(O)c(Cl)c2
IUPAC Name 2,6-dichloro-4-(3-chloro-4-hydroxyphenyl)sulfonylphenol
Molecular Weight (g/mol) 353.61
Formula Weight 351.9131